Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015221.3 | NP_056036.1 | 4 | 698 | ||
XM_006717735.2 | XP_006717798.1 | 4 | 693 | |||
XM_011539559.2 | XP_011537861.1 | 5 | 769 | |||
GenBank mRNA | AL833283.1 | - | 4 | 664 | ||
AY196211.1 | - | 3 | 572 |