Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015235.2 | NP_056050.1 | 1 | 1332 | ||
GenBank mRNA | AB014589.1 | - | 1 | 1080 | ||
AK222574.1 | - | 1 | 1320 | |||
AK314318.1 | - | 1 | 1332 | |||
AL136747.1 | - | 1 | 1328 | |||
AY130299.1 | - | 1 | 1324 | |||
BC028239.1 | - | 1 | 1316 | 5244025 |