Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 3 | 378 | ||
NM_001281711.1 | NP_001268640.1 | 3 | 339 | |||
NM_001281712.1 | NP_001268641.1 | 2 | 158 | |||
NM_015341.4 | NP_056156.2 | 3 | 411 | |||
XM_005263908.3 | XP_005263965.1 | 3 | 351 | |||
XM_006712388.3 | XP_006712451.1 | 3 | 351 | |||
GenBank mRNA | AK291053.1 | - | 3 | 352 | ||
AK299248.1 | - | 3 | 280 | |||
AK303725.1 | - | 2 | 103 | |||
AK308050.1 | - | 3 | 318 | |||
AK310649.1 | - | 3 | 279 | |||
BC024211.2 | - | 3 | 365 | 3504782 | ||
BI559873.1 | - | 3 | 399 | |||
BI560881.1 | - | 3 | 416 | |||
D38553.1 | - | 3 | 341 |