Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319683.1 | NP_001306612.1 | 3 | 195 | ||
NM_053274.2 | NP_444504.1 | 3 | 195 | |||
NR_135089.1 | 3 | 195 | ||||
XM_006710309.2 | XP_006710372.1 | 3 | 173 | |||
XM_011540546.2 | XP_011538848.1 | 5 | 358 | |||
XM_017000137.1 | XP_016855626.1 | 3 | 563 | |||
XM_017000138.1 | XP_016855627.1 | 3 | 563 | |||
XM_017000139.1 | XP_016855628.1 | 3 | 563 | |||
XM_017000141.1 | XP_016855630.1 | 3 | 195 | |||
XM_017000142.1 | XP_016855631.1 | 3 | 173 | |||
XM_017000143.1 | XP_016855632.1 | 3 | 554 | |||
XM_017000144.1 | XP_016855633.1 | 3 | 160 | |||
GenBank mRNA | AJ302735.1 | - | 3 | 161 | ||
AJ347709.1 | - | 3 | 162 | |||
AK295971.1 | - | 3 | 173 | |||
BC001257.1 | - | 3 | 162 | 3454182 | ||
DA729622.1 | - | 3 | 195 | |||
U73704.1 | - | 3 | 173 |