Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 23 | 3474 | ||
NM_001304473.1 | NP_001291402.1 | 24 | 3503 | |||
NM_001304474.1 | NP_001291403.1 | 23 | 3448 | |||
NM_003741.3 | NP_003732.2 | 23 | 3477 | |||
NR_130747.1 | 20 | 3531 | ||||
XM_011513254.1 | XP_011511556.1 | 22 | 3066 | |||
XM_011513256.1 | XP_011511558.1 | 21 | 3486 | |||
XM_017007388.1 | XP_016862877.1 | 22 | 3411 | |||
XM_017007389.1 | XP_016862878.1 | 22 | 3408 | |||
XM_017007390.1 | XP_016862879.1 | 21 | 3147 | |||
XM_017007391.1 | XP_016862880.1 | 21 | 3144 | |||
XM_017007392.1 | XP_016862881.1 | 22 | 3063 | |||
XM_017007394.1 | XP_016862883.1 | 21 | 3199 | |||
GenBank mRNA | AB208947.1 | - | 16 | 4449 | ||
AF038198.1 | - | 9 | 1477 | |||
AF076612.1 | - | 20 | 2769 | |||
AF209928.1 | - | 23 | 3477 | |||
AF209930.1 | - | 24 | 3503 | |||
AF283325.1 | - | 23 | 3448 | |||
AK074147.1 | - | 20 | 3284 | |||
AK293456.1 | - | 23 | 3199 | |||
AY358926.1 | - | 23 | 3364 | |||
BC029879.1 | - | 10 | 1607 |