Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198546.1 | NP_001185475.1 | 3 | 731 | ||
NM_001198547.1 | NP_001185476.1 | 3 | 620 | |||
NM_001198548.1 | NP_001185477.1 | 3 | 731 | |||
NM_001282527.1 | NP_001269456.1 | 4 | 661 | |||
NM_003787.4 | NP_003778.2 | 3 | 731 | |||
XM_006722563.2 | XP_006722626.1 | 4 | 1488 | |||
XM_006722564.2 | XP_006722627.1 | 3 | 473 | |||
XM_006722565.1 | XP_006722628.1 | 4 | 507 | |||
XM_011526237.1 | XP_011524539.1 | 4 | 1485 | |||
XM_011526238.1 | XP_011524540.1 | 3 | 1097 | |||
XM_011526239.2 | XP_011524541.1 | 2 | 489 | |||
XM_017026052.1 | XP_016881541.1 | 2 | 319 | |||
XM_017026053.1 | XP_016881542.1 | 3 | 471 | |||
XM_017026054.1 | XP_016881543.1 | 4 | 599 | |||
XM_017026055.1 | XP_016881544.1 | 4 | 1138 | |||
XM_017026056.1 | XP_016881545.1 | 4 | 392 | |||
XM_017026057.1 | XP_016881546.1 | 3 | 851 | |||
XM_017026058.1 | XP_016881547.1 | 3 | 210 | |||
XM_017026059.1 | XP_016881548.1 | 3 | 2013 | |||
GenBank mRNA | AB017800.1 | - | 3 | 741 | ||
AK091463.1 | - | 4 | 510 | |||
AK297185.1 | - | 3 | 385 | |||
AK299850.1 | - | 3 | 242 | |||
AK312562.1 | - | 4 | 258 | |||
BC000313.2 | - | 3 | 417 | 2821116 | ||
BT006763.1 | - | 2 | 91 | |||
CR456730.1 | - | 2 | 91 |