Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003794.3 | NP_003785.1 | 11 | 1006 | ||
NR_073435.1 | 10 | 884 | ||||
XM_017007414.1 | XP_016862903.1 | 12 | 1094 | |||
GenBank mRNA | AF065485.1 | - | 12 | 964 | ||
AF130078.1 | - | 6 | 373 | |||
AK001835.1 | - | 11 | 968 | |||
AK298972.1 | - | 10 | 851 | |||
BC018762.1 | - | 11 | 982 | 4641014 |