Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271811.1 | NP_001258740.1 | 4 | 362 | ||
NM_004264.4 | NP_004255.2 | 4 | 364 | |||
NR_073463.1 | 3 | 249 | ||||
NR_073464.1 | 4 | 614 | ||||
GenBank mRNA | AK309064.1 | - | 3 | 221 | ||
AK311838.1 | - | 4 | 308 | |||
BC008380.1 | - | 4 | 336 | 4041267 | ||
CR456999.1 | - | 4 | 306 | |||
DB024509.1 | - | 4 | 364 | |||
HY075642.1 | - | 4 | 336 | |||
KU178474.1 | - | 4 | 306 | |||
KU178475.1 | - | 4 | 319 | |||
U46837.1 | - | 4 | 315 |