Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOL4 SIRNA Assays ›
Gene Symbol:
NOL4
Gene Name:
nucleolar protein 4
Gene Aliases:
CT125, HRIHFB2255, NOLP
Chromosome Location:
Chr.18: 33851100 - 34224834 on Build GRCh38
UniGene ID:
Hs.514795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13755
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOL4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198546.1
NP_001185475.1
10
2317
NM_001198547.1
NP_001185476.1
11
2398
NM_001198548.1
NP_001185477.1
9
2203
NM_001198549.1
NP_001185478.1
7
1613
NM_001282527.1
NP_001269456.1
11
2247
NM_003787.4
NP_003778.2
11
2509
XM_006722563.2
XP_006722626.1
12
3266
XM_006722564.2
XP_006722627.1
10
2059
XM_006722565.1
XP_006722628.1
12
2285
XM_011526237.1
XP_011524539.1
12
3263
XM_011526238.1
XP_011524540.1
11
2875
XM_011526239.2
XP_011524541.1
10
2267
XM_017026052.1
XP_016881541.1
10
2097
XM_017026053.1
XP_016881542.1
11
2249
XM_017026054.1
XP_016881543.1
12
2377
XM_017026055.1
XP_016881544.1
11
2724
XM_017026056.1
XP_016881545.1
11
1978
XM_017026057.1
XP_016881546.1
12
2841
XM_017026058.1
XP_016881547.1
11
1988
XM_017026059.1
XP_016881548.1
12
3856
XM_017026060.1
XP_016881549.1
6
1435
GenBank mRNA
AB017800.1
-
11
2518
AK091463.1
-
11
2096
AK296539.1
-
7
1596
BC000313.2
-
9
1889
2821116
Back To Top
More Information
Gene Ontology Categories:
Process(es)
RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0