Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286839.1 | NP_001273768.1 | 6 | 686 | ||
NM_005766.3 | NP_005757.1 | 6 | 865 | |||
XM_011521046.2 | XP_011519348.1 | 6 | 865 | |||
XM_017020312.1 | XP_016875801.1 | 6 | 609 | |||
XM_017020313.1 | XP_016875802.1 | 5 | 1602 | |||
GenBank mRNA | AB008430.1 | - | 6 | 495 | ||
BC041595.1 | - | 6 | 712 | 5443230 | ||
BC071592.1 | - | 6 | 686 | 30342239 |