Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142531.1 | NP_001136003.1 | 10 | 1758 | ||
NM_001142532.1 | NP_001136004.1 | 10 | 2007 | |||
NM_001142533.1 | NP_001136005.1 | 9 | 1895 | |||
NM_001142534.1 | NP_001136006.1 | 9 | 1867 | |||
NM_001252334.1 | NP_001239263.1 | 11 | 2264 | |||
NM_005489.3 | NP_005480.2 | 10 | 1893 | |||
NM_170600.2 | NP_733745.1 | 11 | 2366 | |||
XM_005251639.1 | XP_005251696.1 | 10 | 1874 | |||
XM_011518114.1 | XP_011516416.1 | 11 | 2350 | |||
XM_011518115.1 | XP_011516417.1 | 11 | 2310 | |||
XM_011518117.2 | XP_011516419.1 | 9 | 2986 | |||
XM_017014174.1 | XP_016869663.1 | 9 | 2147 | |||
GenBank mRNA | AF124251.1 | - | 10 | 1805 | ||
AJ420431.1 | - | 4 | 435 | |||
AK056068.1 | - | 9 | 1895 | |||
AK096983.1 | - | 7 | 1188 | |||
AK128287.1 | - | 12 | 2297 | |||
AK291393.1 | - | 9 | 1859 | |||
AK292041.1 | - | 10 | 1821 | |||
AK292718.1 | - | 11 | 2237 | |||
AK294386.1 | - | 10 | 1758 | |||
AY358089.1 | - | 10 | 1875 | |||
AY358440.1 | - | 10 | 1794 | |||
BC027962.1 | - | 10 | 1764 | 5223012 | ||
BC032365.1 | - | 11 | 2241 | 4521962 | ||
BX647905.1 | - | 11 | 2190 | |||
CU454732.1 | - | 3 | 242 | |||
CU455802.1 | - | 3 | 242 |