Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003384.2 | NP_003375.1 | 12, 13 | 1271 | ||
XM_017021624.1 | XP_016877113.1 | 12, 13 | 1306 | |||
XM_017021625.1 | XP_016877114.1 | 12, 13 | 1350 | |||
GenBank mRNA | AB000449.1 | - | 12, 13 | 1240 | ||
AK290110.1 | - | 12, 13 | 1259 | |||
BC103761.1 | - | 12, 13 | 1249 | 5773877 | ||
BC112075.1 | - | 12, 13 | 1239 | 8327543 | ||
BC113510.1 | - | 12, 13 | 1239 | 8322562 |