Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 7 | 1021 | ||
XM_005250060.4 | XP_005250117.1 | 7 | 1057 | |||
XM_011516629.2 | XP_011514931.1 | 7 | 1013 | |||
XM_011516630.2 | XP_011514932.1 | 7 | 914 | |||
XM_011516631.2 | XP_011514933.1 | 7 | 1042 | |||
XM_011516632.2 | XP_011514934.1 | 7 | 940 | |||
GenBank mRNA | AF062536.1 | - | 6 | 795 | ||
AK096163.1 | - | 8 | 909 | |||
AK296909.1 | - | 6 | 746 | |||
AK315023.1 | - | 7 | 1197 | |||
BC125119.1 | - | 7 | 934 | 40118923 | ||
BC125120.1 | - | 7 | 934 | 40118927 | ||
BX537409.1 | - | 7 | 951 | |||
U58087.1 | - | 7 | 723 |