Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122957.2 | NP_001116429.1 | 10 | 1235 | ||
NM_005881.3 | NP_005872.2 | 10 | 1235 | |||
XM_017022859.1 | XP_016878348.1 | 10 | 1148 | |||
GenBank mRNA | AF026548.1 | - | 10 | 1145 | ||
AK130145.1 | - | 10 | 1115 | |||
AK294691.1 | - | 9 | 922 | |||
BC007363.2 | - | 10 | 1118 | 3630050 | ||
CR542093.1 | - | 9 | 872 |