Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282601.1 | NP_001269530.1 | 7 | 1000 | ||
NM_001282602.1 | NP_001269531.1 | 7 | 973 | |||
NM_001282603.1 | NP_001269532.1 | 7 | 964 | |||
NM_001282604.1 | NP_001269533.1 | 9 | 1112 | |||
NM_001282605.1 | NP_001269534.1 | 6 | 910 | |||
NM_006601.6 | NP_006592.3 | 8 | 1063 | |||
NR_104219.1 | 8 | 1972 | ||||
XM_005268576.4 | XP_005268633.1 | 8 | 2653 | |||
XM_006719199.2 | XP_006719262.1 | 7 | 1322 | |||
XM_011537774.2 | XP_011536076.1 | 7 | 1349 | |||
GenBank mRNA | AK098214.1 | - | 8 | 1972 | ||
AK291945.1 | - | 8 | 817 | |||
AK293139.1 | - | 7 | 922 | |||
AK295208.1 | - | 7 | 718 | |||
AK298147.1 | - | 7 | 876 | |||
AK298160.1 | - | 7 | 787 | |||
BC003005.1 | - | 8 | 760 | 2821965 | ||
BC021167.1 | - | 4 | 314 | |||
BU167210.1 | - | 6 | 763 |