Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015092.4 | NP_055907.3 | 17 | 2705 | ||
XM_005255182.3 | XP_005255239.1 | 17 | 2718 | |||
XM_005255183.3 | XP_005255240.1 | 18 | 2776 | |||
XM_005255184.3 | XP_005255241.1 | 16 | 2562 | |||
XM_011545769.2 | XP_011544071.1 | 17 | 2718 | |||
XM_011545770.1 | XP_011544072.1 | 5 | 515 | |||
XM_017023066.1 | XP_016878555.1 | 17 | 2528 | |||
XM_017023067.1 | XP_016878556.1 | 5 | 578 | |||
GenBank mRNA | AB061371.1 | - | 18 | 2657 | ||
AF395444.1 | - | 15 | 2012 | |||
AY014957.1 | - | 5 | 584 |
Gene Symbol: SMG1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027154.1 | 17 | 2725 | |||
GenBank mRNA | AK300040.1 | - | 3 | 200 | ||
BC090929.1 | - | 5 | 634 |
Gene Symbol: SMG1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027155.2 | 17 | 2766 |