Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017775.3 | NP_060245.3 | 2, 3 | 780 | ||
XM_017024801.1 | XP_016880290.1 | 2, 3 | 693 | |||
XM_017024802.1 | XP_016880291.1 | 2, 3 | 693 | |||
GenBank mRNA | AK056878.1 | - | 2, 3 | 692 | ||
AK297783.1 | - | 2, 3 | 336 | |||
BC011698.2 | - | 2, 3 | 319 | |||
BC073796.1 | - | 2, 3 | 323 | |||
BC105128.1 | - | 2, 3 | 780 | 8144131 | ||
BC112107.1 | - | 2, 3 | 780 | 8327575 |