Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018128.4 | NP_060598.3 | 2 | 1081 | ||
GenBank mRNA | AB037822.1 | - | 2 | 1204 | ||
AK026565.1 | - | 2 | 154 | |||
BC110851.1 | - | 2 | 659 | 5786818 | ||
BC126110.1 | - | 2 | 629 | 8991826 |