Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018157.2 | NP_060627.2 | 1, 2 | 217 | ||
XM_005268998.3 | XP_005269055.1 | 1, 2 | 243 | |||
XM_017019548.1 | XP_016875037.1 | 1, 2 | 243 | |||
GenBank mRNA | AK001482.1 | - | 1, 2 | 124 | ||
AY221116.1 | - | 1, 2 | 26 | |||
BC034220.1 | - | 1, 2 | 167 | 5264088 | ||
BC132689.1 | - | 1, 2 | 198 | 40146711 | ||
BC144056.1 | - | 1, 2 | 198 | 9052576 | ||
DA325821.1 | - | 1, 2 | 217 |