Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IQCH SIRNA Assays ›
Gene Symbol:
IQCH
Gene Name:
IQ motif containing H
Gene Aliases:
NYDSP5
Chromosome Location:
Chr.15: 67254772 - 67501804 on Build GRCh38
UniGene ID:
Hs.657894
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140732
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IQCH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031715.2
NP_001026885.1
10
1451
NM_001284347.1
NP_001271276.1
6
988
NM_001284348.1
NP_001271277.1
8
1223
NM_001284349.1
NP_001271278.1
7
1117
NM_001322470.1
NP_001309399.1
7
1117
NM_001322471.1
NP_001309400.1
6
988
NM_001322472.1
NP_001309401.1
6
988
NM_001322473.1
NP_001309402.1
7
1117
NM_001322474.1
NP_001309403.1
7
1117
NM_001322475.1
NP_001309404.1
8
1235
XM_011521907.2
XP_011520209.1
11
1592
XM_011521908.2
XP_011520210.1
10
1451
XM_011521911.2
XP_011520213.1
9
1379
XM_011521912.2
XP_011520214.1
11
1592
XM_011521913.2
XP_011520215.1
11
1593
XM_011521914.2
XP_011520216.1
8
1111
XM_011521915.2
XP_011520217.1
11
1592
XM_011521916.2
XP_011520218.1
7
1030
XM_011521917.2
XP_011520219.1
9
1299
XM_011521918.2
XP_011520220.1
7
1085
XM_017022491.1
XP_016877980.1
11
1592
XM_017022492.1
XP_016877981.1
11
1592
XM_017022493.1
XP_016877982.1
10
1553
XM_017022494.1
XP_016877983.1
11
1592
XM_017022495.1
XP_016877984.1
11
1592
XM_017022496.1
XP_016877985.1
11
1592
XM_017022497.1
XP_016877986.1
11
1592
XM_017022498.1
XP_016877987.1
11
1592
XM_017022499.1
XP_016877988.1
11
1592
XM_017022500.1
XP_016877989.1
8
1258
XM_017022501.1
XP_016877990.1
7
1085
XM_017022502.1
XP_016877991.1
8
1246
XM_017022503.1
XP_016877992.1
6
1011
XM_017022504.1
XP_016877993.1
6
1011
GenBank mRNA
AK022538.1
-
8
1173
AK292478.1
-
10
1403
AY014282.2
-
10
1422
BC031041.1
-
7
1069
BC039292.1
-
6
938
5270043
BC048964.1
-
10
1401
5297681
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0