Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286191.1 | NP_001273120.1 | 6 | 1512 | ||
NM_001286192.1 | NP_001273121.1 | 6 | 1422 | |||
NM_001286195.1 | NP_001273124.1 | 6 | 1389 | |||
NM_001286196.1 | NP_001273125.1 | 6 | 1386 | |||
NM_001286197.1 | NP_001273126.1 | 5 | 1323 | |||
NM_001286198.1 | NP_001273127.1 | 7 | 1679 | |||
NM_022895.2 | NP_075046.1 | 6 | 1419 | |||
NR_104409.1 | 6 | 1432 | ||||
XM_017019829.1 | XP_016875318.1 | 6 | 1503 | |||
GenBank mRNA | AK022510.1 | - | 6 | 1414 | ||
BC014661.1 | - | 6 | 1396 | 4860511 |