Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318834.1 | NP_001305763.1 | 16 | 2242 | ||
NM_032933.5 | NP_116322.3 | 15 | 2127 | |||
NR_134875.1 | 14 | 2078 | ||||
NR_134876.1 | 11 | 1998 | ||||
NR_134877.1 | 14 | 2081 | ||||
XM_011526233.2 | XP_011524535.1 | 16 | 2219 | |||
XM_017026045.1 | XP_016881534.1 | 14 | 2041 | |||
XM_017026049.1 | XP_016881538.1 | 15 | 2123 | |||
XM_017026050.1 | XP_016881539.1 | 15 | 2145 | |||
XM_017026051.1 | XP_016881540.1 | 15 | 2047 | |||
GenBank mRNA | AK123623.1 | - | 11 | 1988 | ||
AK126247.1 | - | 14 | 2070 | |||
BC006280.2 | - | 3 | 1348 | |||
BC054507.1 | - | 9 | 1624 | |||
BC082984.1 | - | 3 | 1475 |