Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025141.3 | NP_079417.2 | 5 | 923 | ||
NM_078474.2 | NP_510883.2 | 6 | 1001 | |||
GenBank mRNA | AF353992.1 | - | 5 | 897 | ||
AK026257.1 | - | 5 | 913 | |||
AK094955.1 | - | 4 | 1333 | |||
AL512689.1 | - | 2 | 1759 | |||
BC006150.1 | - | 6 | 973 | 3051281 | ||
BC008873.2 | - | 5 | 896 | 4137334 |