Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098514.2 | NP_001091984.2 | 8 | 1530 | ||
NM_152459.4 | NP_689672.4 | 8 | 1888 | |||
XM_011522392.2 | XP_011520694.1 | 6 | 1229 | |||
XM_017022972.1 | XP_016878461.1 | 9 | 1451 | |||
GenBank mRNA | AK075242.1 | - | 9 | 1810 | ||
AK300708.1 | - | 8 | 1304 | |||
AY358483.1 | - | 8 | 1853 | |||
BC033681.1 | - | 8 | 1425 | 5190553 |