Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126018.1 | - | 1 | 2715 | ||
BC110528.1 | - | 1 | 2295 | 40034264 | ||
BC110529.1 | - | 1 | 2295 | 40034267 | ||
U79265.1 | - | 1 | 1135 |