Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002773.4 | NP_002764.1 | 4 | 555 | ||
GenBank mRNA | AU142128.1 | - | 4 | 555 | ||
BC001462.2 | - | 4 | 466 | 3138532 | ||
L41351.1 | - | 4 | 519 |