Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNH1 SIRNA Assays ›
Gene Symbol:
RNH1
Gene Name:
ribonuclease/angiogenin inhibitor 1
Gene Aliases:
RAI, RNH
Chromosome Location:
Chr.11: 494512 - 507283 on Build GRCh38
UniGene ID:
Hs.530687
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142493
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002939.3
NP_002930.2
6
1008
NM_203383.1
NP_976317.1
6
940
NM_203384.1
NP_976318.1
5
767
NM_203385.1
NP_976319.1
5
835
NM_203386.2
NP_976320.1
6
898
NM_203387.2
NP_976321.1
6
966
NM_203388.2
NP_976322.1
5
725
NM_203389.2
NP_976323.1
5
793
XM_011520255.1
XP_011518557.1
6
943
XM_011520257.2
XP_011518559.1
6
1191
XM_011520258.2
XP_011518560.1
6
1159
XM_011520259.2
XP_011518561.1
5
708
XM_011520260.2
XP_011518562.1
6
867
XM_011520261.2
XP_011518563.1
6
941
XM_011520262.2
XP_011518564.1
6
1124
XM_011520263.1
XP_011518565.1
5
675
XM_017018106.1
XP_016873595.1
6
1008
GenBank mRNA
AB209196.1
-
4
2258
AL161967.1
-
4
1943
AL534410.3
-
5
781
AL539148.3
-
5
809
BC000677.2
-
5
695
3349381
BC003506.1
-
5
708
2959514
BC011186.1
-
4
493
BC011500.1
-
5
646
4155804
BC014629.2
-
5
695
3029730
BC024037.1
-
5
708
2959514
BC047730.1
-
6
987
5788220
BM562998.1
-
5
766
BQ927358.1
-
5
654
M22414.1
-
5
656
M36717.1
-
6
895
X13973.1
-
6
895
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
negative regulation of catalytic activity
regulation of angiogenesis
protein binding
ribonuclease inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jmz56:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0