Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143166
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
13
2103
NM_001318068.1
NP_001304997.1
13
2198
NM_001318069.1
NP_001304998.1
14
2668
NM_002753.4
NP_002744.1
14
2209
NM_138980.3
NP_620446.1
14
2695
NM_138982.3
NP_620448.1
14
2668
XM_005263129.2
XP_005263186.1
14
2765
XM_005263130.2
XP_005263187.1
14
2161
XM_005263131.2
XP_005263188.1
14
2562
XM_005263135.3
XP_005263192.1
14
2567
XM_006714268.2
XP_006714331.1
14
2435
XM_006714269.2
XP_006714332.1
14
2567
XM_011532117.2
XP_011530419.1
14
2268
XM_011532118.2
XP_011530420.1
14
2361
XM_011532120.2
XP_011530422.1
14
2331
XM_011532121.2
XP_011530423.1
14
2197
XM_017008419.1
XP_016863908.1
15
2458
XM_017008420.1
XP_016863909.1
14
2892
XM_017008421.1
XP_016863910.1
14
2488
XM_017008422.1
XP_016863911.1
11
1916
XM_017008423.1
XP_016863912.1
14
2180
XM_017008424.1
XP_016863913.1
14
2515
XM_017008425.1
XP_016863914.1
14
2188
XM_017008426.1
XP_016863915.1
14
2516
XM_017008427.1
XP_016863916.1
14
2589
XM_017008428.1
XP_016863917.1
14
2331
XM_017008429.1
XP_016863918.1
14
2897
XM_017008430.1
XP_016863919.1
14
2275
XM_017008431.1
XP_016863920.1
14
2493
XM_017008432.1
XP_016863921.1
14
2166
XM_017008433.1
XP_016863922.1
13
2517
XM_017008434.1
XP_016863923.1
14
2550
XM_017008435.1
XP_016863924.1
14
2440
XM_017008436.1
XP_016863925.1
14
2924
XM_017008437.1
XP_016863926.1
14
2202
XM_017008438.1
XP_016863927.1
14
2193
XM_017008439.1
XP_016863928.1
14
2521
XM_017008440.1
XP_016863929.1
14
2594
XM_017008441.1
XP_016863930.1
13
2522
XM_017008442.1
XP_016863931.1
16
2910
XM_017008443.1
XP_016863932.1
16
2561
XM_017008444.1
XP_016863933.1
16
2883
XM_017008445.1
XP_016863934.1
13
2392
XM_017008446.1
XP_016863935.1
11
3084
XM_017008447.1
XP_016863936.1
13
2530
XM_017008448.1
XP_016863937.1
13
2503
XM_017008449.1
XP_016863938.1
13
2530
XM_017008450.1
XP_016863939.1
13
2503
XM_017008451.1
XP_016863940.1
13
2334
XM_017008452.1
XP_016863941.1
13
2535
GenBank mRNA
AK057723.1
-
9
1995
AK091104.1
-
6
1934
AK124791.1
-
14
2920
AK225974.1
-
14
2249
BC022492.1
-
14
2501
BC035057.1
-
14
2513
5261221
BC051731.1
-
14
2314
4838495
BC065516.1
-
9
1665
5721954
U07620.1
-
14
2210
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0