Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099855.1 | NP_001093325.1 | 11 | 1216 | ||
NM_001171876.1 | NP_001165347.1 | 11 | 1216 | |||
NM_001171877.1 | NP_001165348.1 | 7 | 1081 | |||
NM_001171878.1 | NP_001165349.1 | 8 | 1198 | |||
NM_001171879.1 | NP_001165350.1 | 8 | 1198 | |||
NM_005369.4 | NP_005360.3 | 8 | 1198 | |||
XM_005262413.4 | XP_005262470.1 | 12 | 1364 | |||
XM_011531339.2 | XP_011529641.1 | 12 | 1423 | |||
XM_017029529.1 | XP_016885018.1 | 11 | 1185 | |||
XM_017029530.1 | XP_016885019.1 | 11 | 1185 | |||
XM_017029531.1 | XP_016885020.1 | 11 | 1118 | |||
XM_017029532.1 | XP_016885021.1 | 8 | 1275 | |||
GenBank mRNA | AB085901.1 | - | 8 | 875 | ||
AB085902.1 | - | 8 | 875 | |||
AK296488.1 | - | 7 | 1081 | |||
AK296529.1 | - | 11 | 1316 | |||
AK302957.1 | - | 12 | 1164 | |||
AK313900.1 | - | 8 | 1058 | |||
AK316416.1 | - | 11 | 1204 | |||
AL117234.1 | - | 10 | 1099 | |||
X12556.1 | - | 8 | 1049 |