Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144466
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
12
2294
NM_021038.4
NP_066368.2
10
3025
NM_207292.2
NP_997175.1
9
2989
NM_207293.1
NP_997176.1
9
2418
NM_207294.1
NP_997177.1
7
2160
NM_207295.1
NP_997178.1
8
2196
NM_207296.1
NP_997179.1
8
2274
NM_207297.1
NP_997180.1
8
2305
XM_005247457.4
XP_005247514.1
12
2647
XM_005247458.4
XP_005247515.1
12
3901
XM_005247460.4
XP_005247517.1
12
2610
XM_005247461.4
XP_005247518.1
11
2611
XM_005247462.4
XP_005247519.1
11
2593
XM_005247463.4
XP_005247520.1
10
2557
XM_005247464.4
XP_005247521.1
11
2705
XM_005247465.4
XP_005247522.1
11
2552
XM_005247466.4
XP_005247523.1
10
2646
XM_005247467.4
XP_005247524.1
10
2516
XM_005247468.4
XP_005247525.1
9
2462
XM_005247471.4
XP_005247528.1
9
2528
XM_005247474.3
XP_005247531.1
10
1565
XM_005247475.3
XP_005247532.1
11
1888
XM_005247476.4
XP_005247533.1
9
1601
XM_005247477.4
XP_005247534.1
10
1654
XM_006713639.1
XP_006713702.1
12
2629
XM_011512843.2
XP_011511145.1
11
2443
XM_011512845.2
XP_011511147.1
10
2389
XM_011512846.2
XP_011511148.1
9
2353
XM_011512847.2
XP_011511149.1
9
2352
XM_011512848.2
XP_011511150.1
12
2251
XM_011512849.2
XP_011511151.1
8
2260
XM_017006423.1
XP_016861912.1
11
2898
XM_017006424.1
XP_016861913.1
11
2725
XM_017006425.1
XP_016861914.1
10
2691
XM_017006426.1
XP_016861915.1
10
2689
XM_017006427.1
XP_016861916.1
10
2818
XM_017006428.1
XP_016861917.1
10
2482
XM_017006429.1
XP_016861918.1
10
2565
XM_017006430.1
XP_016861919.1
9
2593
XM_017006431.1
XP_016861920.1
9
2670
XM_017006432.1
XP_016861921.1
10
2587
XM_017006433.1
XP_016861922.1
9
2551
XM_017006434.1
XP_016861923.1
9
2557
XM_017006435.1
XP_016861924.1
11
1695
XM_017006436.1
XP_016861925.1
9
3659
XM_017006437.1
XP_016861926.1
8
2678
XM_017006438.1
XP_016861927.1
9
2314
XM_017006439.1
XP_016861928.1
10
1834
XM_017006440.1
XP_016861929.1
9
1568
XM_017006441.1
XP_016861930.1
8
1565
XM_017006442.1
XP_016861931.1
8
1668
XM_017006443.1
XP_016861932.1
7
1581
GenBank mRNA
AB007888.1
-
9
2989
AF255334.1
-
7
1882
AK297133.1
-
6
1080
AK307420.1
-
7
1120
BC043493.1
-
10
2530
6044671
BC050535.1
-
12
1815
6042311
BX537491.1
-
2
2098
CR749454.1
-
4
653
Y13829.1
-
9
2140
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001253710, NM_001253711, NM_001253713, NM_001253708, NM_020007, and others.
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0