Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GOLGB1 SIRNA Assays ›
Gene Symbol:
GOLGB1
Gene Name:
golgin B1
Gene Aliases:
GCP, GCP372, GOLIM1
Chromosome Location:
Chr.3: 121663199 - 121749908 on Build GRCh38
UniGene ID:
Hs.213389
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145095
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GOLGB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256486.1
NP_001243415.1
4, 5
536
NM_001256487.1
NP_001243416.1
4, 5
564
NM_001256488.1
NP_001243417.1
4, 5
564
NM_004487.4
NP_004478.3
4, 5
536
XM_005247371.4
XP_005247428.1
4, 5
996
XM_005247372.4
XP_005247429.1
4, 5
572
XM_005247373.1
XP_005247430.1
4, 5
511
XM_006713587.1
XP_006713650.1
4, 5
511
XM_006713588.1
XP_006713651.1
4, 5
511
XM_006713589.1
XP_006713652.1
4, 5
511
XM_006713590.1
XP_006713653.1
4, 5
511
XM_006713591.1
XP_006713654.1
4, 5
511
XM_011512699.2
XP_011511001.1
4, 5
463
XM_017006189.1
XP_016861678.1
4, 5
996
XM_017006190.1
XP_016861679.1
4, 5
1166
XM_017006191.1
XP_016861680.1
4, 5
996
XM_017006192.1
XP_016861681.1
4, 5
572
XM_017006193.1
XP_016861682.1
4, 5
572
XM_017006194.1
XP_016861683.1
4, 5
572
GenBank mRNA
AB371588.1
-
4, 5
509
AB593126.1
-
4, 5
538
AK296903.1
-
2, 3
235
AK298986.1
-
2, 3
210
AK300437.1
-
4, 5
510
BC065301.1
-
4, 5
435
BP320001.1
-
4, 5
537
D25542.1
-
4, 5
519
X75304.1
-
4, 5
524
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
ER to Golgi vesicle-mediated transport
Golgi organization
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0