Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005282.2 | NP_005273.1 | 2 | 2929 | ||
XM_017026607.1 | XP_016882096.1 | 2 | 3517 | |||
XM_017026608.1 | XP_016882097.1 | 2 | 2838 | |||
GenBank mRNA | BC038450.1 | - | 2 | 2916 |