Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPH3 SIRNA Assays ›
Gene Symbol:
HNRNPH3
Gene Name:
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases:
2H9, HNRPH3
Chromosome Location:
Chr.10: 68331174 - 68343196 on Build GRCh38
UniGene ID:
Hs.643472
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145327
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPH3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322434.1
NP_001309363.1
9
1187
NM_001322436.1
NP_001309365.1
9
1149
NM_001322437.1
NP_001309366.1
9
1252
NM_001322438.1
NP_001309367.1
9
1079
NM_001322439.1
NP_001309368.1
9
1142
NM_001322440.1
NP_001309369.1
9
1104
NM_001322441.1
NP_001309370.1
9
1207
NM_001322442.1
NP_001309371.1
9
1169
NM_001322443.1
NP_001309372.1
9
1150
NM_001322444.1
NP_001309373.1
7
844
NM_001322445.1
NP_001309374.1
8
985
NM_001322446.1
NP_001309375.1
8
1029
NM_001322447.1
NP_001309376.1
8
1010
NM_001322448.1
NP_001309377.1
8
1048
NM_001322449.1
NP_001309378.1
8
1113
NM_001322450.1
NP_001309379.1
8
1030
NM_001322451.1
NP_001309380.1
8
965
NM_001322452.1
NP_001309381.1
8
984
NM_001322453.1
NP_001309382.1
8
1003
NM_012207.2
NP_036339.1
9
1168
NM_021644.3
NP_067676.2
9
1123
XM_011539742.1
XP_011538044.1
7
815
XM_017016168.1
XP_016871657.1
9
1598
XM_017016169.1
XP_016871658.1
8
1459
GenBank mRNA
AF132361.1
-
9
1012
AF132362.1
-
7
1069
AF132363.1
-
4
1204
AF132364.1
-
3
1042
AK091411.1
-
7
3040
AK223441.1
-
9
1120
AK291547.1
-
9
1075
AK295316.1
-
7
759
AL117395.1
-
6
2390
BC004511.2
-
6
524
BC039824.1
-
9
1059
6061550
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA processing
RNA splicing
epithelial cell differentiation
nucleotide binding
RNA binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0