Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282175.1 | NP_001269104.1 | 6 | 681 | ||
NM_001282176.1 | NP_001269105.1 | 6 | 705 | |||
NM_001294.3 | NP_001285.1 | 6 | 832 | |||
GenBank mRNA | AF037339.1 | - | 6 | 675 | ||
AK074935.1 | - | 6 | 676 | |||
AK075262.1 | - | 6 | 675 | |||
AK223553.1 | - | 6 | 676 | |||
AK293314.1 | - | 6 | 697 | |||
AK304480.1 | - | 6 | 664 | |||
AK310826.1 | - | 6 | 705 | |||
AK316094.1 | - | 6 | 701 | |||
BC004865.2 | - | 6 | 711 | 3840991 | ||
BC012359.1 | - | 6 | 665 | 4685026 | ||
BI916245.1 | - | 6 | 678 | |||
BT007262.1 | - | 6 | 660 | |||
KU177986.1 | - | 6 | 660 | |||
KU177987.1 | - | 6 | 660 |