Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319039.1 | NP_001305968.1 | 2 | 486 | ||
NM_001319041.1 | NP_001305970.1 | 2 | 486 | |||
NM_001319042.1 | NP_001305971.1 | 2 | 227 | |||
NM_001446.4 | NP_001437.1 | 2 | 486 | |||
GenBank mRNA | AB208815.1 | - | 2 | 233 | ||
AI879148.1 | - | 2 | 529 | |||
AJ002962.1 | - | 2 | 242 | |||
AK307272.1 | - | 2 | 246 | |||
AK311867.1 | - | 2 | 245 | |||
AL512688.1 | - | 2 | 202 | |||
BC012299.1 | - | 2 | 352 | 4707233 | ||
BI860090.1 | - | 2 | 460 | |||
BX461253.2 | - | 2 | 207 | |||
CR457057.1 | - | 2 | 166 | |||
D50373.1 | - | 2 | 218 | |||
D88648.1 | - | 2 | 217 | |||
KM111186.1 | - | 2 | 227 | |||
U51338.1 | - | 2 | 221 | |||
U81235.1 | - | 2 | 181 |