Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004436.2 | NP_004427.1 | 2, 3 | 341 | ||
NM_207044.1 | NP_996927.1 | 2, 3 | 341 | |||
NM_207046.1 | NP_996929.1 | 2, 3 | 205 | |||
NM_207047.1 | NP_996930.1 | 2, 3 | 205 | |||
GenBank mRNA | AF067170.1 | - | 2, 3 | 298 | ||
AF157509.1 | - | 2, 3 | 295 | |||
AF157510.1 | - | 2, 3 | 295 | |||
AK001981.1 | - | 2, 3 | 315 | |||
AK290064.1 | - | 2, 3 | 188 | |||
AL711949.1 | - | 2, 3 | 344 | |||
BC000436.2 | - | 2, 3 | 289 | 2820528 | ||
BC004461.1 | - | 2, 3 | 289 | 2820528 | ||
BC068544.1 | - | 2, 3 | 306 | 5273162 | ||
BE799496.1 | - | 2, 3 | 225 | |||
BQ883543.1 | - | 2, 3 | 303 | |||
CR457130.1 | - | 2, 3 | 189 | |||
CR536578.1 | - | 2, 3 | 189 | |||
T78564.1 | - | 2, 3 | 221 | |||
X99906.1 | - | 2, 3 | 314 |