Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004427.3 | NP_004418.2 | 2 | 464 | ||
NM_198040.2 | NP_932157.1 | 9 | 1771 | |||
XM_005270570.1 | XP_005270627.1 | 2 | 400 | |||
XM_017000513.1 | XP_016856002.1 | 10 | 2270 | |||
XM_017000514.1 | XP_016856003.1 | 10 | 1882 | |||
XM_017000515.1 | XP_016856004.1 | 10 | 1975 | |||
XM_017000516.1 | XP_016856005.1 | 10 | 2204 | |||
XM_017000517.1 | XP_016856006.1 | 10 | 1816 | |||
XM_017000518.1 | XP_016856007.1 | 10 | 1909 | |||
XM_017000519.1 | XP_016856008.1 | 10 | 2279 | |||
XM_017000520.1 | XP_016856009.1 | 9 | 1729 | |||
XM_017000521.1 | XP_016856010.1 | 9 | 2148 | |||
XM_017000522.1 | XP_016856011.1 | 10 | 2309 | |||
GenBank mRNA | AJ419231.1 | - | 9 | 1771 | ||
AK056429.1 | - | 3 | 701 | |||
AK292905.1 | - | 10 | 2209 | |||
AK294426.1 | - | 4 | 1049 | |||
BC018602.1 | - | 2 | 470 | |||
BC028396.1 | - | 2 | 435 | |||
BC029269.1 | - | 2 | 306 | |||
BC068573.1 | - | 2 | 358 | |||
BC092492.1 | - | 2 | 358 | |||
BC110863.1 | - | 2 | 357 | |||
BC130630.1 | - | 8 | 1687 | 40146661 | ||
BC144121.1 | - | 8 | 1687 | 9052641 | ||
CD109667.1 | - | 2 | 481 | |||
U89278.1 | - | 2 | 459 |