Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146880
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
9
1465
NM_001166006.1
NP_001159478.1
9
1538
NM_001166007.1
NP_001159479.1
9
1448
NM_004437.3
NP_004428.1
10
1528
NM_203342.2
NP_976217.1
10
1545
NM_203343.2
NP_976218.1
8
1360
XM_005245753.1
XP_005245810.1
9
1540
XM_005245757.1
XP_005245814.1
9
1540
XM_005245760.1
XP_005245817.1
9
1540
XM_005245761.1
XP_005245818.1
9
1540
XM_005245763.1
XP_005245820.1
9
1540
XM_005245764.1
XP_005245821.1
9
1540
XM_005245765.1
XP_005245822.1
9
1540
XM_005245768.1
XP_005245825.1
9
1540
XM_005245769.1
XP_005245826.1
9
1540
XM_005245770.1
XP_005245827.1
9
1540
XM_005245772.4
XP_005245829.1
9
1540
XM_005245773.4
XP_005245830.1
9
1540
XM_005245774.1
XP_005245831.1
9
1540
XM_006710434.1
XP_006710497.1
9
1540
XM_006710439.1
XP_006710502.1
9
1540
XM_011540956.1
XP_011539258.1
9
1540
XM_011540957.1
XP_011539259.1
9
1540
XM_011540958.1
XP_011539260.1
9
1540
XM_011540959.1
XP_011539261.1
9
1540
XM_011540960.1
XP_011539262.1
9
1540
XM_011540961.1
XP_011539263.1
9
1540
XM_011540962.1
XP_011539264.1
9
1540
XM_011540963.2
XP_011539265.1
9
1540
XM_011540964.1
XP_011539266.1
9
1436
XM_011540965.2
XP_011539267.1
9
1540
XM_017000581.1
XP_016856070.1
8
1435
XM_017000582.1
XP_016856071.1
8
1435
XM_017000583.1
XP_016856072.1
9
1540
XM_017000584.1
XP_016856073.1
9
1831
XM_017000585.1
XP_016856074.1
9
1828
XM_017000586.1
XP_016856075.1
9
1540
XM_017000587.1
XP_016856076.1
9
1540
XM_017000588.1
XP_016856077.1
9
1540
XM_017000589.1
XP_016856078.1
8
1435
XM_017000590.1
XP_016856079.1
8
1435
XM_017000591.1
XP_016856080.1
9
1540
XM_017000592.1
XP_016856081.1
9
1540
XM_017000593.1
XP_016856082.1
9
1436
XM_017000594.1
XP_016856083.1
9
1436
XM_017000595.1
XP_016856084.1
9
1436
XM_017000596.1
XP_016856085.1
9
1436
XM_017000597.1
XP_016856086.1
9
1436
XM_017000598.1
XP_016856087.1
9
1436
XM_017000599.1
XP_016856088.1
9
1436
XM_017000600.1
XP_016856089.1
9
1436
XM_017000601.1
XP_016856090.1
9
1436
XM_017000602.1
XP_016856091.1
9
1436
XM_017000603.1
XP_016856092.1
9
1436
XM_017000604.1
XP_016856093.1
9
1436
GenBank mRNA
AB209649.1
-
9
1448
AF156225.1
-
8
1338
AK310655.1
-
8
1376
BC039079.1
-
9
1535
5167140
BC096104.1
-
6
754
BC096105.1
-
8
1072
40001729
BC113841.1
-
8
1072
BC114450.1
-
8
1072
J03796.1
-
8
1279
M14993.1
-
10
1509
M61733.1
-
10
1526
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0