Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004450.2 | NP_004441.1 | 4 | 539 | ||
GenBank mRNA | AK130850.1 | - | 4 | 525 | ||
BC014301.2 | - | 4 | 522 | 3507241 | ||
BC071709.1 | - | 4 | 486 | 5503738 | ||
D85758.1 | - | 4 | 542 | |||
U66871.1 | - | 4 | 539 |