Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK3 SIRNA Assays ›
Gene Symbol:
ANK3
Gene Name:
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases:
ANKYRIN-G, MRT37
Chromosome Location:
Chr.10: 60026298 - 60733526 on Build GRCh38
UniGene ID:
Hs.499725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147782
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001149.3
NP_001140.2
21
4582
NM_001204403.1
NP_001191332.1
44
6720
NM_001204404.1
NP_001191333.1
44
6963
NM_001320874.1
NP_001307803.1
43
6977
NM_020987.4
NP_066267.2
44
14507
XM_005269715.3
XP_005269772.1
44
6864
XM_006717796.3
XP_006717859.1
47
9483
XM_006717802.3
XP_006717865.1
46
6903
XM_011539708.2
XP_011538010.1
47
9486
XM_011539709.2
XP_011538011.1
46
9177
XM_017016110.1
XP_016871599.1
47
14721
XM_017016111.1
XP_016871600.1
46
14709
XM_017016112.1
XP_016871601.1
47
14706
XM_017016113.1
XP_016871602.1
46
14694
XM_017016114.1
XP_016871603.1
47
14820
XM_017016115.1
XP_016871604.1
47
14550
XM_017016116.1
XP_016871605.1
46
14412
XM_017016117.1
XP_016871606.1
46
14409
XM_017016118.1
XP_016871607.1
47
13380
XM_017016119.1
XP_016871608.1
46
13071
XM_017016120.1
XP_016871609.1
46
13068
XM_017016121.1
XP_016871610.1
45
13044
XM_017016122.1
XP_016871611.1
44
9393
XM_017016123.1
XP_016871612.1
46
9174
XM_017016124.1
XP_016871613.1
45
9150
XM_017016125.1
XP_016871614.1
45
9147
XM_017016126.1
XP_016871615.1
44
9045
XM_017016127.1
XP_016871616.1
43
9018
XM_017016128.1
XP_016871617.1
45
6876
XM_017016129.1
XP_016871618.1
45
6873
XM_017016130.1
XP_016871619.1
43
6807
XM_017016131.1
XP_016871620.1
43
6795
XM_017016132.1
XP_016871621.1
43
6774
XM_017016133.1
XP_016871622.1
43
6909
XM_017016134.1
XP_016871623.1
43
6744
XM_017016135.1
XP_016871624.1
43
6873
XM_017016136.1
XP_016871625.1
45
6594
XM_017016137.1
XP_016871626.1
44
6567
XM_017016138.1
XP_016871627.1
44
6564
XM_017016139.1
XP_016871628.1
42
6501
XM_017016140.1
XP_016871629.1
42
6498
XM_017016141.1
XP_016871630.1
43
6462
XM_017016142.1
XP_016871631.1
42
6450
XM_017016143.1
XP_016871632.1
42
6435
XM_017016144.1
XP_016871633.1
42
6432
XM_017016145.1
XP_016871634.1
41
6423
XM_017016146.1
XP_016871635.1
41
6420
XM_017016147.1
XP_016871636.1
42
6414
XM_017016148.1
XP_016871637.1
42
6317
XM_017016149.1
XP_016871638.1
41
6519
XM_017016150.1
XP_016871639.1
44
14346
GenBank mRNA
AK092361.1
-
11
2307
AK128784.1
-
3
1225
BX537917.1
-
44
6591
BX648574.1
-
44
6685
U13616.1
-
44
14253
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
ER to Golgi vesicle-mediated transport
plasma membrane organization
cytoskeletal anchoring at plasma membrane
signal transduction
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of gene expression
positive regulation of cell communication by electrical coupling
positive regulation of sodium ion transport
magnesium ion homeostasis
neuronal action potential
positive regulation of homotypic cell-cell adhesion
Golgi to plasma membrane protein transport
regulation of potassium ion transport
establishment of protein localization
positive regulation of membrane potential
cellular response to magnesium ion
membrane assembly
protein localization to plasma membrane
maintenance of protein location in plasma membrane
protein targeting to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of delayed rectifier potassium channel activity
positive regulation of sodium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
cytoskeletal adaptor activity
spectrin binding
protein binding, bridging
ion channel binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-kd7lp:80/10.239.160.160:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0