Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004547.5 | NP_004538.2 | 2, 3 | 413 | ||
GenBank mRNA | AF044957.1 | - | 2, 3 | 342 | ||
BC000855.1 | - | 2, 3 | 385 | 3459215 | ||
BC070285.1 | - | 2, 3 | 352 | 6714676 | ||
BC146885.1 | - | 2, 3 | 384 | 9056813 | ||
BC146887.1 | - | 2, 3 | 384 | 9056815 | ||
BF978138.1 | - | 412 |