Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168331.1 | NP_001161803.1 | 2 | 1392 | ||
NM_004547.5 | NP_004538.2 | 3 | 442 | |||
GenBank mRNA | AF044957.1 | - | 3 | 371 | ||
BC000855.1 | - | 3 | 414 | 3459215 | ||
BC070285.1 | - | 3 | 381 | 6714676 | ||
BC146885.1 | - | 3 | 413 | 9056813 | ||
BC146887.1 | - | 3 | 413 | 9056815 | ||
CB215975.1 | - | 1 | 350 |