Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173664.5 | NP_775935.1 | 4 | 1402 | ||
XM_011534531.2 | XP_011532833.1 | 4 | 896 |