Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122665.3 | NP_001116137.1 | 4 | 557 | ||
NM_001302552.2 | NP_001289481.1 | 3 | 205 | |||
NM_001324195.1 | NP_001311124.1 | 3 | 290 | |||
NM_004660.4 | NP_004651.2 | 3 | 290 | |||
NR_136716.1 | 3 | 290 | ||||
NR_136717.1 | 3 | 290 | ||||
NR_136718.1 | 3 | 290 | ||||
NR_136719.1 | 3 | 290 | ||||
NR_136720.1 | 3 | 290 | ||||
NR_136721.1 | 3 | 218 | ||||
NR_136722.1 | 3 | 205 | ||||
NR_136723.1 | 3 | 205 | ||||
NR_136724.1 | 3 | 205 | ||||
GenBank mRNA | AK296366.1 | - | 3 | 194 | ||
AK302172.1 | - | 3 | 205 | |||
AK303176.1 | - | 3 | 243 | |||
AK303638.1 | - | 3 | 253 | |||
AK310382.1 | - | 3 | 195 | |||
BC034942.2 | - | 4 | 448 | 4826417 | ||
DC394594.1 | - | 3 | 290 |