Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258248.1 | NP_001245177.1 | 2 | 2137 | ||
NM_199262.2 | NP_954871.1 | 2 | 2126 | |||
XM_006722115.3 | XP_006722178.1 | 2 | 2089 | |||
GenBank mRNA | AK127707.1 | - | 2 | 2179 |