Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205850.2 | NP_995322.1 | 2 | 250 | ||
GenBank mRNA | AF348468.1 | - | 2 | 250 | ||
BC113628.1 | - | 2 | 205 | 8322680 | ||
BC113630.1 | - | 2 | 205 | 8322682 | ||
BC143950.1 | - | 2 | 205 | 9052468 | ||
DQ665306.1 | - | 2 | 198 |