Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206998.1 | NP_996881.1 | 2 | 251 | ||
GenBank mRNA | AY236538.1 | - | 2 | 218 | ||
AY359052.1 | - | 2 | 251 | |||
BC130639.1 | - | 2 | 226 | 40146670 | ||
BC130645.1 | - | 2 | 226 | 40146676 |