Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207444.2 | NP_997327.1 | 2 | 730 | ||
GenBank mRNA | AK093014.1 | - | 2 | 730 |