Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008946.4 | NP_032972.3 | 5 | 542 | ||
GenBank mRNA | AK078437.1 | - | 5 | 514 | ||
AK132042.1 | - | 5 | 525 | |||
AK167123.1 | - | 5 | 514 | |||
AK167227.1 | - | 5 | 510 | |||
BC013897.1 | - | 5 | 502 | 4038758 | ||
U13393.1 | - | 5 | 463 |