Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044744.1 | NP_001038209.2 | 5, 6 | 540 | ||
NM_008097.2 | NP_032123.3 | 4, 5 | 597 | |||
GenBank mRNA | AK078866.1 | - | 2, 3 | 150 | ||
AK165406.1 | - | 4, 5 | 423 | |||
AU080952.1 | - | 5, 6 | 397 | |||
BC061158.1 | - | 4, 5 | 427 | 30314273 | ||
U18992.1 | - | 4, 5 | 378 |